Learn More
Electron Microscopy Sciences Wehnelt Aperture 200 µm Hole
Supplier:  Electron Microscopy Sciences 49020TA
Encompass_Preferred
Made from 0.004" tantalum foil. Replaceable tantalum apertures for some microscopes (Amray and Philips). They improve the performance of the electron gun.
Catalog No. 50-980-821