Learn More
IMEB, Inc Warthin Rapid Starry Stain Kit. Contains 8 Oz Bottle/Solutions Of: Stabilizer Silver Nitrate, Gelatinn, Hydroquinone Solution, 3- 2 Oz. Staining Containers, and Storage Tray.
Supplier: IMEB, Inc K7408
Rapid Wathin Starry Stain kit is used to detect Cat Scratch, Legionella, illustrates an exception development for Helicobacter. The result is black to brown development against a light yellow background. Not recommended for Spirochetes. Includes: 1- 8 oz. Bottle Stabilizer Silver Nitrate solution, 1- 8 oz. Bottle Gelatin solution, 1- 8 oz. Bottle Hydroquinone solution, 3- 2 oz. Staining containers and storage tray.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.