Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Immunodiagnostic Products B·R·A·H·M·S PCT Reagent
Ortho-Clinical Diagnostics™ B•R•A•H•M•S PCT is a biomarker for early bacterial infection diagnosis and antibiotic stewardship
Supplier: Ortho-Clinical Diagnostics™ 6197735
This item is not returnable.
View return policy
Description
Test delivers:
- High sensitivity and specificity for bacterial infection enables therapeutic decision making
- Results that are ready to be delivered to a clinicians with 96.5% First Pass Yield (without user intervention)
- Analytical performance: LOD at 0.007 ng/mL, LOQ (20% CV, observed) at 0.013 ng/mL
- Fast turnaround time: 24 minutes to first result
- Reliable solution: Not impacted by biotin interference
Indicated as an aid to be used in conjunction with clinical evaluation for:
- Risk assessment of critically ill patients on their first day of ICU admission for progression to severe sepsis and septic shock
- Assessing the cumulative 28-day risk of all-cause mortality for patients diagnosed with severe sepsis or septic shock in the ICU or when obtained in the emergency department or other medical wards prior to ICU admission, using a change in PCT level over time
- Decision making on antibiotic therapy for patients with suspected or confirmed lower respiratory tract infections (LRTI) defined as community-acquired pneumonia (CAP), acute bronchitis, and acute exacerbation of chronic obstructive pulmonary disease (AECOPD) – in an inpatient setting or an emergency department
- Decision making on antibiotic discontinuation for patients with suspected or confirmed sepsis
Indicated for use with the VITROS™ ECi/ECiQ/3600 Immunodiagnostic Systems and the VITROS™ 5600/XT 7600 Integrated Systems.
Specifications
Reagent | |
0.030 to 100 ng/mL (0.030 to 100 μg/L) | |
24 min. |
100 Tests | |
30 μL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction