Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radians VisionAid™ Basic Glasses Sideshields
Click to view available options
Clear
Description
- Dust and nuisance protection
- Easy on and off
- Made with acetate
- Color: clear
Specifications
Color | Clear |
Product Type | Sideshields |
For Use With (Equipment) | Fit all types of eyewear |
For Use With (Application) | Dust and nuisance protection |
Material | Acetate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction