Learn More
Creative Biomart Influenza A virus H1N1 (Beijing) Antigen
Supplier:  Creative Biomart VAC0268
Encompass_Preferred
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Beijing/262/95. The Influenza Virus was purified by Ultracentrifugation with 10-40 % sucrose gradient.
Catalog No. 50-208-5108
Due to product restrictions, please to purchase or view availability for this product.