Learn More
Chemglass Life Sciences 50mL Peptide Synthesis Vessel, Solid Phase, Luer, Medium Frit, GL 25 Thread
SDP
Supplier:  Chemglass Life Sciences CG186203
Encompass_Preferred
50mL Peptide Synthesis Vessel, Solid Phase, Luer, Medium Frit, GL 25 Thread. Similar to CG-1860, but with the addition of a Luer inner joint below the 2mm PTFE stopcock. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. 50-873-360