Learn More
Perkin Elmer US LLC UV Quartz SUPRASIL Cylindrical Cell with PTFE Stopper - 10 mm, Qty. 1
Supplier: Perkin Elmer US LLC B0631095
Cylindrical cells have an outside diameter of 22 mm and an inside diameter of 19 mm. A package contains one cell and two windows. UV Quartz SUPRASIL cells are made from a quartz of the highest purity and homogeneity and have a spectral range of 200 - 2,500 nm. These cells are identified by a blue PerkinElmer logo.