Learn More
Parker Ultraviolet (UV) Germacidal Lamp
SDP
Lamp operates at 254 nm, and features four high intensity UV-C lamps to sterilize the air by deactivating viruses, bacteria, and spores.
Supplier:  Parker 201066
Catalog No. 14-950-436
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Ultraviolet germicidal lamp operates at a wavelength of 254 nm
  • Four high intensity UV-C lamps sterilize the air of bioaerosols
  • UV-C uses ultraviolet light to deactivate viruses, bacteria, and spores
  • Replacement pack includes all 4 lamps required for the CRYSTAL-AIRE™uv System
Specifications
Specifications
Deactivating viruses, bacteria, and spores
4 High Intensity UV-C Lamps
UV Lamp
Product Suggestions
Product Suggestions