Learn More
MAPA™ Ultrane 524 ESD Glove
Catalog Number 19046605
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy
Lightweight, PU coated, general purpose glove with electrostatic discharge properties and touchscreen capabilities.
  • ESD Properties
  • Touchscreen Capabilites
  • PU Coated
  • Lightweight
  • Comfort
  • Breathability
  • Silicon Free
  • Washable
Coating Material Polyamide, Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Hand Specific
Latex Free No
Material Elastomer, PU
Size 11
Gender Unisex
Glove Cuff Style Continuous Knit
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Wing
Length (Metric) 27 cm