Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSG Series General Purpose Laboratory Refrigerators

Designed for your everyday laboratory needs

Supplier:  Thermo Scientific™ TSG30RSGA

Request Bulk Order

 View more versions of this product

Catalog No. TSG30RSGA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Thermo Scientific™ TSG Series general-purpose laboratory refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2° to 10°C controlled environment.

  • Featuring a wide range of cabinet sizes from 12 cu. ft. to 72 cu. ft.
  • Whisper-quiet operation
  • ENERGY STAR certifiedTemperature, door ajar*, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Warranty: varies by country, please contact us for details
    *All models except TSG45RPLA and TSG45RSLA come equipped with door ajar alarm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG Series general purpose laboratory refrigerator
764 L
+5°C
Glass
4 Full
45 kg
Painted
cULus, ENERGYSTAR, Prop 65
1 Rear access port
Adaptive Defrost
Key Lock
32x30.75x81 in.(813x781x2057 mm)
NEMA 5-15 P
115 V 60 Hz
Energy efficient
175 kg
27 cu. ft.
2°C to 10°C
R290
1
100 lb.
Gray-painted
Stainless Steel
Digital Microprocessor
2 years parts and labor, plus 3-year compressor warranty
Alarm
28.5x27x61.75 in. (724x686x1569 mm)
3 A
115 V
60 Hz
386 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit