Learn More
United States Biological Corporation TRIS BASE ULTRAPURE 1KG
Supplier: United States Biological Corporation T86001KG
Tris is one of the most widely used buffers in molecular biology and cell culture due to its low toxicity, stability and buffering capacity. Purity: ≥99.9% Appearance: White, crystalline powder pH (1M): 10.5-11.5 pKa: 8.0-8.4 Absorbance (260nm): ≤0.06 Absorbance (280nm): ≤0.05 Absorbance (290nm): ≤0.2 Solubility (1M): Colorless, clear, complete Loss on Drying: ≤0.2% Melting Point: 168-172°C Heavy Metals (Pb): ≤0.0001% Arsenic: ≤0.0001% Calcium: ≤0.0001% Copper: ≤0.0001% Iron: ≤0.0005% Magnesium: ≤0.0001% Insoluble Matter: ≤0.005% Residue on Ignition: ≤0.1% RNase: DNase: Protease: Meets specifications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.