Learn More
Cole-Parmer Wika S-10 Pressure Transmitter, 0 to 5000 psi
Supplier: Cole-Parmer 6844698
The industrial design provides a rugged transmitter suitable for many applications, including test equipment, press control, compressor control, pump protection, hydraulics, pneumatics and numerous other process and control applications.
Applications: Hydraulics and pneumatics Test equipment Pump and compressor control Liquid level measurement