Learn More
Cole-Parmer Wika S-10 Pressure Transmitter, 30 inHg to 0 inHg
Supplier:  Cole-Parmer 6844706
Encompass_Preferred
The industrial design provides a rugged transmitter suitable for many applications, including test equipment, press control, compressor control, pump protection, hydraulics, pneumatics and numerous other process and control applications.
Applications: Hydraulics and pneumatics Test equipment Pump and compressor control Liquid level measurement
Catalog No. 50-992-329