Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Doppler Ultrasonic Transducer for 1"" and Larger Pipe
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3261566
Use these doppler flowmeters measure the flow of liquids in pipes from 1/4" and larger. Each flowmeter requires a transducer (32615-66 or -68) and a monitor (32615-60, -62, or -64).|The transducers send flow data via RG59 coax cable to the monitor, and the monitor¿s software automatically adjusts filtering parameters and gain settings to ensure reliable readings are ensured over a wide range of liquid and reflector conditions. Easily configure the monitor using the front panel keypad. A two-line backlit LCD displays rate and accumulated flow values in user selected measuring units. Models 32615-62 and -64 have a 4 to 20 mA input and output for datalogging or recording. A 4 to 20 mA input must be supplied for a 4 to 20 mA output. The monitor is housed in a NEMA 4X (IP-66) enclosure. The NEMA 6 (IP67) rating has the ability to work with temperatures up to 400¿F (200¿C).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.