Learn More
Cole-Parmer Ashcroft K1100PSI Thin Film Pressure Transmitter, 0 to 100 psi, 1.0%
Supplier: Cole-Parmer 6844036
This transmitter offers the benefits of polysilicon thin film performance at affordable prices. Using chemical vapor deposition methods polysilicon strain gauges are molecularly bonded to a metal diaphragm. This sensor, combined with the latest surface mount electronics promote superior performance in a durable package.