Learn More
Midwest Scientific TPP Tubespin Bioreactor, 50ml, 60/case
SDP
Supplier:  Midwest Scientific TP8705060
Encompass
Features: Working Volume 1-35Ml *Easy To Start Small Suspension Cell Cultures From Frozen *Ergonomic Use Of Bacterial And Yeast Cultures -- Static And Shaking Conical Bottom 0.2um Filter Membrane *Sterile Gas Exchange 5 Openings *(A, B, C, D, E) Of Different Size Above The Gas Permeable, Sterile Ptfe Filter Of The Screw Cap *Openings Can Be Sealed To Adjust Gas Exchange According To Need Fits In Standard 15Ml Centrifuge Rotors *Saves Costs Of Culture Start-Ups In Erlenmyer Flasks And Then Transfer To Conical Tubes *Culture And Spin In A Single Tube. 60/case
Catalog No. NC0664085