Learn More
Malvern Panalytical Tip ,100Ul Plunger(Pk/5)
Supplier: Malvern Panalytical TIP130010005
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Replacement plunger tips for MicroCal PEAQ-ITC, PEAQ-ITC Automated, ITC200 and Auto-ITC200.