Learn More
Terra Universal Tool Kit
Tools for working with, stripping, and coating wire
Supplier: Terra Universal 930035
Includes:
- Hook for dressing of gold wire and removal of bond wire
- Fine point for applying small amounts of epoxy, removing foreign materials in small areas, and dressing gold wire
- Medium point for applying epoxy in small areas, removing foreign materials, and tuning gold ribbon
- Chisel for removal of bond wire, dressing gold pads prior to bonding, and removal of large foreign material
Description
- Color-coded handles for easy identification
- Accurate manipulation of wire bonds, gold ribbon, and aids in dressing and tuning
- Laying down and removing epoxy
- Removing foreign material in small areas
Specifications
Universal Electrical Work Tool Kit | |
Ideal for accurate manipulation of wire bonds, Gold ribbon, aids in tuning and indispensable Laying down and removing Epoxy, and removing foreign material | |
Blue hook, white fine point, black medium point, gray chisel |