Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Sysmex™ CA Reaction Tubes
For use with Sysmex™ CA System Hemostatis Analyzer
Supplier: Siemens Healthineers 10874002
Description
Specifications
Reaction Tubes | |
Sysmex™ CA Systems | |
3000 Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction