Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Syringes Fitted with Luer Lock Valve
Feature an interference fit between the plunger tip and the barrel to improve the syringe's gas sealing properties
Supplier: MilliporeSigma™ Supelco™ 509620
Description
- Maximum inertness for liquid or gas samples
- Suitable for manual dispenser/diluter applications
- High accuracy of dispensed volume
- Syringes are autoclavable when plunger is removed
Specifications
± 1% | |
Borosilicate Glass (Barrel), PTFE (Plunger) | |
Syringe | |
1 Ea. | |
Luer Lock, Kel-F Luer Cones |
Autoclavable | |
Pre-fitted with Luer Lock Valve | |
10 MDR-VLLMA-GT | |
10 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction