Learn More
Synthware™ Buchner Filter Funnels
GREENER_CHOICE
Used to filter particulates from a variety of substances
Supplier:  Synthware™ F609600M
 View more versions of this product
Catalog No. 31-501-537
Add to Cart
Description
Description
  • Sealed in glass fritted disc of listed size and porosity
  • Porosity: XC-Extra Coarse, C-Coarse, M-Medium, F-Fine
  • Not recommended for filtration where pressure would exceed 15 psi
Specifications
Specifications
Buchner filter funnel
600 mL
90mm
Medium
Product Suggestions
Product Suggestions