Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Flammables Safety Can Storage Cabinets With Handles
GREENER_CHOICESCI_ED

Keep dangerous liquids safely organized and segregated. Justrite™ Sure-Grip™ EX Flammables Safety Can Storage Cabinets With Handles feature flush mounted handles, two keys for security and have the ability to be padlocked. Dual vents with flame arresters allow hazardous vapers to be piped away.

$1762.00 - $3387.00

Specifications

Color Yellow
Material Steel (CR)
Product Type Can storage cabinet
Width (English) Exterior 43 in.
For Use With (Application) Flammable liquid storage and protection
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (D x W x H) Interior No. of Shelves Certifications/Compliance Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (D x W x H) Interior No. of Shelves Certifications/Compliance Door Style Price Quantity & Availability  
S28708
GREENER_CHOICESCI_ED
Accessories View Documents
Justrite™
893020/EMD
14.56 x 39.5 x 39.62 in. (37 x 100.3 x 100.6 cm) 1 FM, NFPA, OSHA, U (NFPA) Self-close
Each for $1,762.00
Add to Cart
 
S28750
GREENER_CHOICESCI_ED
View Documents
Justrite™
899020/EMD
30.34 x 39.5 x 60.5 in. (77.1 x 100.3 x 153.7 cm) 2 FM, NFPA, OSHA, U (NFPA) Self-close
Each for $3,387.00
Add to Cart
 
S28749
SCI_ED
View Documents
Justrite™
899000/EMD
30.34 x 39.5 x 60.5 in. (77.1 x 100.3 x 153.7 cm) 2 FM, NFPA, OSHA Manual-close Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product
N/A
Description

Description

  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements
Specifications

Specifications

Yellow
Can storage cabinet
Flammable liquid storage and protection
2
39.5 in.
100.3 cm
Steel (CR)
43 in.
U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and more!
Epoxy, Polyester
109 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit