Learn More
Metro™ Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit, 74 in., Metroseal Gray Epoxy PROMO
Super Erecta 4-Shelf Industrial Wire Shelving Starter Unit consist of four posts and four Super Erecta shelves.
$729.00 - $1356.00
Specifications
Autoclavable Not Autoclavable
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
For Use With (Equipment) Super Erecta Shelving
Height (English) 74 in.
Load Bearing Capacity (English) 2000 lb.
View More Specs
Products 24
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
13-460-418PM
View Documents
Metro™
SE182474K4S4PM
18 x 24 in.
Each for $729.00
Add to Cart
13-460-419PM
View Documents
Metro™
SE183074K4S4PM
18 x 30 in.
Each for $757.00
Add to Cart
13-460-404PM
View Documents
Metro™
SE183674K4S4PM
18 x 36 in.
Each for $757.00
Add to Cart
13-460-402PM
View Documents
Metro™
SE184274K4S4PM
18 x 42 in.
Each for $886.00
Add to Cart
13-460-444PM
View Documents
Metro™
SE184874K4S4PM
18 x 48 in.
Each for $862.00
Add to Cart
13-460-408PM
View Documents
Metro™
SE185474K4S4PM
18 x 54 in.
Each for $1,005.00
Add to Cart
13-460-421PM
View Documents
Metro™
SE186074K4S4PM
18 x 60 in.
Each for $978.00
Add to Cart
13-460-432PM
View Documents
Metro™
SE187274K4S4PM
18 x 72 in.
Each for $1,096.00
Add to Cart
13-460-409PM
View Documents
Metro™
SE212474K4S4PM
21 x 24 in.
Each for $810.00
Add to Cart
13-460-430PM
View Documents
Metro™
SE213074K4S4PM
21 x 30 in.
Each for $828.00
Add to Cart
13-460-436PM
View Documents
Metro™
SE213674K4S4PM
21 x 36 in.
Each for $828.00
Add to Cart
13-460-440PM
missing translation for 'viewDocuments'
Metro™
SE214274K4S4PM
21 x 42 in.
Each for $939.00
Add to Cart
13-460-431PM
missing translation for 'viewDocuments'
Metro™
SE214874K4S4PM
21 x 48 in.
Each for $939.00
Add to Cart
13-460-400PM
missing translation for 'viewDocuments'
Metro™
SE215474K4S4PM
21 x 54 in.
Each for $1,061.00
Add to Cart
13-460-427PM
missing translation for 'viewDocuments'
Metro™
SE216074K4S4PM
21 x 60 in.
Each for $1,061.00
Add to Cart
13-460-433PM
missing translation for 'viewDocuments'
Metro™
SE217274K4S4PM
21 x 72 in.
Each for $1,233.00
Add to Cart
13-460-425PM
missing translation for 'viewDocuments'
Metro™
SE242474K4S4PM
24 x 24 in.
Each for $877.00
Add to Cart
13-460-410PM
missing translation for 'viewDocuments'
Metro™
SE243074K4S4PM
24 x 30 in.
Each for $891.00
Add to Cart
13-460-437PM
missing translation for 'viewDocuments'
Metro™
SE243674K4S4PM
24 x 36 in.
Each for $866.00
Add to Cart
13-460-442PM
missing translation for 'viewDocuments'
Metro™
SE244274K4S4PM
24 x 42 in.
Each for $1,010.00
Add to Cart
13-460-412PM
missing translation for 'viewDocuments'
Metro™
SE244874K4S4PM
24 x 48 in.
Each for $1,010.00
Add to Cart
13-460-439PM
missing translation for 'viewDocuments'
Metro™
SE245474K4S4PM
24 x 54 in.
Each for $1,155.00
Add to Cart
13-460-414PM
missing translation for 'viewDocuments'
Metro™
SE246074K4S4PM
24 x 60 in.
Each for $1,155.00
Add to Cart
13-460-434PM
missing translation for 'viewDocuments'
Metro™
SE247274K4S4PM
24 x 72 in.
Each for $1,356.00
Add to Cart
Description
Description
  • Warm gray look that blends with most environments
  • Superior rust protection than chrome wire shelving
  • Built-in Microban antimicrobial product protection
  • 12-year corrosion resistance warranty
Specifications
Specifications
Not Autoclavable
Super Erecta Shelving
2000 lb.
4
Starter Unit
Epoxy-coated Steel
All Laboratory and Clean Room Non-Autoclavable Applications
74 in.
907 kg
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. Metroseal shelves and posts also carry a twelve (12) year warranty against rust formation and corrosion.
Gray