Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ ABZ+Plus Supelguard Cartridge Kit, 5 μm Particle Size

Offer both high deactivation and unique selectivity.

$443.00

Specifications

Quantity 1 Kit
Particle Size 5 μm
Length (Metric) 2 cm
Pore Size 120 Å
Type Guard Cartridge Kit
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity & Availability  
11-100-3889
View Documents
MilliporeSigma™ Supelco™
59604
2.1 mm
Each for $443.00
Add to Cart
 
11-100-3887
View Documents
MilliporeSigma™ Supelco™
59534U
4 mm
Each for $443.00
Add to Cart
 
Description

Description

SUPELCOSIL™ ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte's functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions. Suitable for L60 per USP.
Specifications

Specifications

1 Kit
2 cm
Guard Cartridge Kit
Reversed Phase
SUPELCOSIL Supelguard
5 μm
120 Å
1 cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules (10 mm i.d guard requires the purchase of 567499-U)
HPLC
SDS
Documents

Documents

Product Certifications

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit