Learn More
Metrohm USA Storage solution for all combined pH glass electrodes with reference electrolyte c(KCl) = 3 mol/L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Metrohm USA 62323000
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Storage solution for all combined pH glass electrodes with reference electrolyte c(KCl) = 3 mol/L, 250 mL. Long-term storage in KCl damages the glass as it results in ever-increasing response times. The patented 6.2323.000 Storage solution for combined pH glass electrodes has been developed to solve this problem. If a combined pH glass electrode is kept in the Storage solution then the performance of the glass membrane does not change regarding response time and the alkali error. If c(KCl) = 3 mol/L is additionally used as the reference electrolyte then the optimized composition of the Storage solution guarantees that the pH glass electrode is always ready for immediate use. Conditioning before the measurement is not necessary, no matter how long the electrode has been stored.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.