Learn More
Hach Company Still, Automatic, 120 Vac, 50/60 HZ
Supplier: Hach Company 2615900
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
The easy-to-use Barnstead/Thermolyne FI-Streem II 2S Glass Still provides low volumes (2.0 liters per hour) of high-quality distilled water. The still removes both inorganic and organic impurities. Patented vapor trap eliminates carry-over of vapor-borne water droplets which may contain impurities. The still's space-saving design allows for wall or benchmounting. The still has fully automatic controls, a level sensor, and dual feed capability. CSA approved. Pressure: 10-80 psi. Dimensions: 79 x 26 x 32.5 cm (31.5 x 10.5 x 13"). Shipping weight: 15 kg (33 lbs).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.