Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
Supplier:  Troemner™ 30390965
Catalog No. 19-216-365
Description
Description
  • One- and two-piece construction, depending on weight
  • Includes certificate of accuracy
  • Includes case with latch
  • Made in the USA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Weight Set with No Certificate
Stainless Steel
1.6 in.
Class 7
Plastic
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg
Product Suggestions
Product Suggestions