Learn More
Electron Microscopy Sciences Water Bath Stabilizing Ring for 125-500 ml flasks, 237 gm, 48 mm
Supplier:  Electron Microscopy Sciences 64436
Encompass_Preferred
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Weight: 237 gm
I.D.: 48 mm
Fits: 125-500ml flasks
Catalog No. 50-289-81