Learn More
Teledyne ISCO, Inc Stainless Steel pH and Temperature Probe
Supplier: Teledyne ISCO, Inc 609004126
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Stainless steel double junction combination pH probe with built-in exposed temperature probe and 25 ft. (7.62 m) cable.