Learn More
3M™ Speedglas™ Welding Filter G5-01VC
Click to view available options
Black
Description
- Dark shade options include 5, 8-14.
- Natural Color technology which makes the filter view seem lighter, more detailed and colorful
- Auto-ON/OFF
- ADFs are Bluetooth enabled
- Utilizing the 3M Connected Equipment app, users get access to additional functionalities, statistics and documentation.
- Selecting just the right welding filter settings for each task reduces eye strain and may help improve the quality of the weld.
- Switching time from light to dark in 0.1 millisecond, the sensitivity of arc detection can be adjusted for increased swithching reliability.
Specifications
Lens Tint | Shade 8 to 14 |
Product Type | G5 Series Filter |
Model | 46-0000-30IVC |