Learn More
Cone Instruments SONO ULTRASOUND WIPES 50/PK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Cone Instruments CN17297
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum qty of 4 per supplier requirements.
Designed by Ultrasound Engineers and Sonographers, SONO Ultrasound Wipes is the only disinfectant wipe that uses an EPA approved Proprietary Patented Solution which kills 47 pathogens including MRSA, HIV, Staph and has been tested to prevent damage to your equipment and transducers saving you thousands in repair costs. There is no need for multiple cleaning products in your ultrasound department. SONO Ultrasound Wipes cleans, sanitizes, disinfects and deodorizes everything it touches.Each wipe is 7" x 10" Won't damage cables or lenses Safe to handle without gloves Safe on touch screens and system consoles Now effective against the 2013 influenza A virus (H7N9) This product is phosphate, bleach, and ammonia free.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.