Learn More
World Precision Instrument Small FORT Force Transducers-FORT25
Supplier: World Precision Instrument FORT25
The FORT Force Transducers are reliable tools for high precision force measurement. Using balanced semiconductor strain gauges, the force transducer produces linear output voltage versus applied force input with very little deflection. It is a temperature-compensated, full-bridge configuration with four high sensitivity semiconductor strain gauges. It has a broad dynamic measuring range and a very high sensitivity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.