Learn More
Scientific Digital Imaging Signa Gel, 250g, 12/pk
Supplier: Scientific Digital Imaging PE1525
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Highly conductive, multi-purpose electrolyte meets all the standards of the ideal saline electrode gel. Recommended for ECG, defibrillation, biofeedback and EMG. 250g (8.5oz.) tube, 12 per box