Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Histology Stains and Reagents - Schiff's Reagent
For the demonstration of lymphocytes and mucopolysaccharides
Supplier: Sigma Diagnostics™ 3952016/DUP
Description
- For use in Periodic Acid Schiff (PAS) Sigma Procedure No. 395
Applications:
For in vitro diagnostic use
Specifications
Colorless | |
For use with blood, bone marrow, tissue touch preparations or routine tissue sections. | |
500mL |
Solution | |
Reagent | |
2° to 8°C |
Safety and Handling
ShelfLife : Expiry date on the label
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction