Learn More
SURE TECH DIAGNOSTIC ASSOC INC ST 301-B Sickle Cell Test Kit (50 tests)
Supplier: SURE TECH DIAGNOSTIC ASSOC INC ST 301B
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sickle Cell Test Kits. Simple turbidity test for detection of hemoglobin S in anticoagulated blood. 50 test kit