Learn More
Sigma Aldrich Fine Chemicals Biosciences Serum Replacement 2 (50×), 100 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences S9388100ML
Defined, multi-purpose serum replacement with additional components for cells requiring higher concentrations of serum than 10%. Contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and bovine insulin. Does not contain growth factors, steroid hormones, glucocorticoids, cell adhesion factors, detectable Ig and mitogens. May be used with anchorage-dependent and suspension cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.