Learn More
Metrohm USA Septum 16 mm, Set of 5 pieces
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Metrohm USA 61448020
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Septum 16 mm, 5 pieces. Height 2mm; Material: Silicone rubber; Outer diameter 16mm