Learn More
Electron Microscopy Sciences Mini SEM Clamp 10mm, 15mm dia.
Supplier: Electron Microscopy Sciences 7595519
Aluminum Mini SEM clamp on a 10 mm, 15 mm diameter M4 cylinder mount. Convenient for holding flat or thin specimens such as silicon chips, wires, foil and small tubes. Screw is M2 x 3 mm, pan head screw, brass.