Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen VIP-Fixative™ Non-precipitating, 10% Formalin, Neutral pH

Compatible with and replaces traditional 10% NBF. Eliminates routine warm water flushes, and salt accumulation in all tissue processors. Neutralex™ compatible for Down-the-Drain disposal.

$227.00 - $232.00

Specifications

Color Clear, Water White
Compatibility Compatible with Aqueous and Alcoholic Solutions
Content And Storage Room temperature
Concentration Neutral Buffered 10% Formalin
For Use With (Equipment) All Tissue Processors
View More Specs

Products 2
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
23-730-581
View Documents
Scigen
5990
3.8 L
Case of 4 for $232.00
Edge
Add to Cart
 
23-730-582
View Documents
Scigen
5991
19 L
Each for $227.00
Add to Cart
 
Description

Description

  • Introduced in 2001, VIP-Fixative has been marketed under several well-known brands worldwide for use in automatic tissue processors as a non-precipitating formalin fixative
  • VIP-Fixative formula will not form solids (precipitate) in tissue processors which can eliminate the need for routine warm water flushing by preventing precipitate damage to the rotary valve.
  • Clear, colorless solution free of particles, neutral buffered 10% formalin based, for use in histological applications of fixation and storage of tissue specimens, soluble is dehydrant reagents
  • Improves cellular details and improves cell wall rigidity resulting in easier cutting and uniform sections
  • Non-sterile containers marked with a lot code and expiration date, 4 year shelf life
  • Formalin waste designed to be treated in bulk using Neutralex
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Specifications

Clear, Water White
Room temperature
All Tissue Processors
Formalin
Compatible with Aqueous and Alcoholic Solutions
Neutral Buffered 10% Formalin
Formalin
Tissue processor, to prevent instrument damage from solids with no impact on specimen integrity
Safety and Handling

Safety and Handling

ShelfLife : Stable

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit