Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saxagliptin hydrochloride
DPPIV/CD26 Inhibitors
Specifications
CAS | 709031-78-7 |
---|---|
Synonym | (1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride |
Molecular Formula | C18H25N3O2.HCl |
Formula Weight | 351.87 |
Purity | 98% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
65-075-0
|
R&D Systems
6507/50 |
50 mg |
Each for $567.24
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
65-071-0
|
R&D Systems
6507/10 |
10 mg |
Each for $136.88
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
High affinity DPP-IV inhibitor (Ki = 0.6 nM). Inhibits plasma DPP-IV (87%). Elicits enhanced glucose clearance in Zuckerfa/fa rats.Specifications
709031-78-7 | |
C18H25N3O2.HCl | |
98% |
(1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride | |
351.87 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title