Learn More
Honeywell Salisbury™ AS1200 Arc Flash Protection Face Shield
Weight–balancing shield stows in a balanced, compact position, centered over the top of the hard hat when not needed
Supplier: Honeywell Salisbury™ AS1200
This item is not returnable.
View return policy
Description
- The shield/window is designed to be easily replaced without tools
- The faceshield unit includes ventilation ribs that allow a natural convection air flow through the top of the face shield
- Reduces fogging of the visor, lowers CO2 levels and improves comfort for the user
Specifications
Yellow, Orange |