Learn More
Tecan SaliCap Set 500 sets
Supplier:  Tecan RE69995.
Encompass_Preferred
Sampling tubes for collection of saliva samples to be used in the IBL Progesterone Luminescence Immunoassay. Collection tubes (max. 2 mL volume each) 500 sets
Catalog No. 50-130-6186