Learn More
TerraCycle™ Safety Equipment & Protective Gear Zero Waste Box™
Recycle Safety Equipment & Protective Gear. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Specifications
Color | Brown, Green, White |
---|---|
Autoclavable | Not Autoclavable |
Material | Cardboard |
Product Type | Recycling Box |
Alert:
These boxes are not suitable for medical waste. This includes any red bag waste (i.e. material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.) The zero waste box system does not accept household items that may be considered hazardous waste. Do not ship dripping packages.
Not available in Hawaii, Alaska, and Puerto Rico.
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Capacity (Metric) | Price | Quantity & Availability | ||||
S475652
|
TerraCycle™
CS58S |
11 x 11 x 20 in. | 39.66 L |
Each for $178.00
|
N/A | ||||
S475653
|
TerraCycle™
CS58M |
11 x 11 x 40 in. | 79.31 L |
Each for $341.00
|
N/A | ||||
S475654
|
TerraCycle™
CS58L |
15 x 15 x 37 in. | 136.42 L |
Each for $559.00
|
N/A | ||||
Description
- WHAT YOU CAN RECYCLE
- Use this Zero Waste Box to recycle all brands and sizes of disposable safety equipment and protective gear.
- This includes:
- Ear plugs
- Beard nets
- Hair nets
- Gloves
- Safety glasses
- Disposable Garments
- Masks
- Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
Specifications
Brown, Green, White | |
Not Autoclavable | |
Cardboard | |
Recycling Box |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.