Learn More
Hettich Centrifuges ROTOFIX32A,(4)Cyto,BioSeal,Package
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Hettich Centrifuges 32ABIOSEAL
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Hettich CYTOLOGY BIO-SEAL PACKAGE ROTOFIX 32A, (1) Centrifuge, (1) Horizontal Rotor, (4) Cyto Buckets, (4) Slide carriers, (8) One funnel chamber 1mL, (8) One funnel chamber 2mL, (4) Lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years