Learn More
Electron Microscopy Sciences Rinzl Disposable Plastic Micro Slide (A
Supplier:  Electron Microscopy Sciences 7189012
Encompass_Preferred
Disposable plastic micro slides made from optical clear vinyl with a refractive index of glass. Ideal for wet mount, urines, etc. Economical, they can be used and discarded. Will not break, scratch, or cut fingers. Not auto-fluorescent. Thickness 0.0180 to 0.022" or 0.46 to 0.56 mm. Available standard size 3 x 1" and packaged 144 per box.
Catalog No. 50-949-084