Learn More
BD Rhapsody™ Enhanced Cartridge Reagent Kit
The purpose of this kit is to provide the necessary reagents needed to prepare the BD Rhapsody™ Cartridges, extract RNA and antibody oligos bound to the cells, and capture them on BD Rhapsody™ Cell Capture Beads.
Supplier:  BD 664887
Catalog No. BDB664887
This item is not returnable. View return policy
Description
Description
  • Provides the necessary reagents needed to prepare the BD Rhapsody™ Cartridges, extract RNA and antibody oligos bound to the cells, and capture them on BD Rhapsody™ Cell Capture Beads
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Product Suggestions
Product Suggestions