Learn More
Chemglass Life Sciences 10L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve, with CG-1969-M-01 Manifold System
Supplier: Chemglass Life Sciences CG1965610M
10L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve, with CG-1969-M-01 Manifold System. Complete glass reactor system on our NEW open air, auto-center stand. PLEASE NOTE: REACTOR STAND OVERALL HEIGHT WITH MOTOR AND CLEARANCE BELOW VALVE, ARE CALCULATED FOR ALL CHEMGLASS STIRRER BEARINGS, INCLUDING THE CG-2077-M MECHANICAL SEAL STIR BEARING.Support Frame with Oversized Rugged S.S. Uprights and Heavy Duty PTFE Coated Aluminum Fittings. Perfluoro O-Rings On 60mm and 200mm Flanges. S.S. Inlet/Outlet Adapters Available In M16 x 1 and M30 x 1.5 Allow Connection To Cooling/Heating Circulator.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.