Learn More
Ace Glass, Inc. 1000mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, 0-8mm valve and 28/15 ball joint bottom outlet, 28/15 ball joint inlet/outlet, complete with addition funnel, condenser, manifold & stirring assemblies
SDP
Supplier:  Ace Glass, Inc. 6439248
Encompass_Preferred
Request Bulk Order
Complete, one-piece 1000mL jacketed pressure reactor is designed to perform low to moderate positive pressure reactions, synthesis, and catalysis. Flask has a #25 center neck, (3) #15 side necks, and (1) #7 side neck. Flask bottom outlet is 0-8mm 28/15 ball joint. Jacket inlet/outlet has a 28/15 o-ring ball joint. Rated 45 PSI at 100 degrees C. Reactor system includes addition funnel, condenser, manifold, and stirring assemblies.
Catalog No. 50-972-960