Learn More
Ace Glass, Inc. 1000mL, one-piece pressure reactor with #25 center, (3) #15, (1) #7 side necks, 0-8mm valve and 28/15 ball joint bottom outlet, complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies
Supplier: Ace Glass, Inc. 6437238
1000mL one-piece pressure reactor featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet. Reactor is complete with an addition funnel, condenser, heating mantle, pressure manifold, stir shaft and temperature controller. Unit will perform positive pressure reactions up to 45psi at 100 C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.