Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers RAPIDLab™ 1265 Blood Gas Analyzer

RAPIDLab™ 1200 systems are intended for in vitro diagnostic use by healthcare professionals in the quantitative testing of human blood. The 1265 can determine pH, pCO2, pO2, Na+, K+, Ca++, Cl-, glucose, lactate, tHb, nBili, FO2Hb, FCOHb, FMetHb, FHHb parameters.

Supplier:  Siemens Healthineers 10321852

Request Bulk Order

Catalog No. 23-044-847


Add to Cart
This item is not returnable. View return policy

Description

Description

  • For medium- to high-volume critical care testing sites
  • Full test menu from a small blood sample
  • Self-contained, easily replaceable reagent and wash cartridges
  • Easy-to-use system features three simple steps
  • Enhanced system security with McAfee Embedded Control, firewall and verified system connection
  • Immediate results available in just 1 minute
  • Environmental Requirements:
    • Temperature: 15 to 32°C
    • Humidity: 5 to 90% noncondensing
    • Barometric Pressure: 523 to 800 mmHg (69.7 to 106.7 kPa)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USB (three ports), RS232 port, bar-code scanner port and RJ10/100 Base T Ethernet port
∼60 sec.
McAfee embedded control, firewall, verified system connection and encrypted patient data transfer via USB
Rating: 150 VA, Voltage: 100/240 VAC, Freq: 50/60 Hz
Heparinized Whole Blood
One point every 30 minutes to a maximum of 60 minutes; Full calibration every 8 hours
pH only: 95 μL; CO-oximetry only: 100 μL; Full menu: 175 μL
TUV-listed to IEC/EN 61010, 61010-2-101, 61010-2-081, CB Test Certificate, certified to CSA and UL
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit